Learn More
Eleven Components Phenol Standard solution, 2000μg/mL of each, Fisher Chemical™
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Brand: Fisher Chemical F115881
Code : 04
Additional Details : Weight : 0.00130kg Transport : UN number : 1593 Chem class : 6.1 Pack group : III
Specifications
Eleven Components Standard Solution | |
Ampoule | |
Organic Multi-component Standard Solution | |
4-Chloro-3-methylphenol (CAS:59-50-7), 2-Chlorophenol (CAS:95-57-8), 2,4- Dichlorophenol (CAS:120-83-2), 2,4-Dimethylphenol (CAS:105-67-9), 2,4-Dinitrophenol (CAS:51-28 -5), 2-Methyl-4,6-dinitrophenol (CAS:534-52-1), 2-Nitrophenol (CAS:88-75-5), 4-Nitrophenol (CAS:100-02-7), Pentachlorophenol (CAS:87-86-5), Phenol (CAS:108-95-2), 2,4,6-Trichlorophenol (CAS:88-06-2) |
2000μg/mL | |
1 mL | |
1593 | |
Dichloromethane |
Safety and Handling
- Standard solution 11 Components in dichloromethane
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Carcinogenicity Category 2
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H351-Suspected of causing cancer.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P202-Do not handle until all safety precautions have been read and understood.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P330-Rinse mouth.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P501-Dispose of contents/container to .