missing translation for 'onlineSavingsMsg'
Learn More

Phenylmagnesium bromide, 3M in ether, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Catalog Number 15414885
Click to view available options
Quantity:
100 mL
100-58-3
C6H5BrMg
181.32
MFCD00000038
ANRQGKOBLBYXFM-UHFFFAOYSA-M
66852
Br[Mg]C1=CC=CC=C1
100-58-3
C6H5BrMg
181.32
MFCD00000038
ANRQGKOBLBYXFM-UHFFFAOYSA-M
66852
Br[Mg]C1=CC=CC=C1

Phenylmagnesium bromide is an important reagent called as Grignard reagent used in the synthesis of organic and oraganometallic reactions. It is used to prepare phenyl aryl sulphides, sulphoxides, and sulphones. It acts as a strong acid and Lewis acid and undergoes addition reaction with many unsaturated functional groups. It is involved in the synthesis of end-functionalized regioregular poly(3-alkylthiophene)s, 1,3,3-trimethyl-6-phenyl-2-oxabicyclo[2.2.2]octan-6-ol, o-substituted benzophenones and 6-benzyl-1,3,3-trimethyl-2-oxabicyclo[2.2.2]octan-6-ol. In addition to this, it is used for the monalkylation of primary amines to get secondary amines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY
Color Yellow to Brown
Density 1.14 g/mL
Odor Ether-like
Packaging ChemSeal™ bottles
Quantity 100 mL
UN Number UN3399
Sensitivity Air, light, and moisture sensitive; May form a Precipitate
Synonym phenylmagnesium bromide, bromophenylmagnesium, magnesium, bromophenyl, phmgbr, phenylmagnesiumbromide, bromo phenyl magnesium, phenyl magnesium bromide, phenylmagnesium bromide solution, 3.0 m in diethyl ether, bromomagnesiobenzene, phenyhnagnesium bromide, grignard reagent
Solubility Information Miscible with tetrahydrofuran.
IUPAC Name magnesium;benzene;bromide
Formula Weight 181.31
Concentration or Composition (by Analyte or Components) 3M in ether
Physical Form Liquid
Chemical Name or Material Phenylmagnesium bromide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'