missing translation for 'onlineSavingsMsg'
Learn More

Phloroglucinol, 99+%, anhydrous, Thermo Scientific Chemicals

Catalog Number 11994671
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
108-73-6
C6H6O3
126.1
QCDYQQDYXPDABM-UHFFFAOYSA-N
phloroglucinol, 1,3,5-benzenetriol, 1,3,5-trihydroxybenzene, phloroglucin, phloroglucine, spasfon-lyoc, s-trihydroxybenzene, benzene-s-triol, 5-hydroxyresorcinol, benzene, trihydroxy
359
CHEBI:16204
benzene-1,3,5-triol
C1=C(C=C(C=C1O)O)O
108-73-6
C6H6O3
126.1
QCDYQQDYXPDABM-UHFFFAOYSA-N
phloroglucinol, 1,3,5-benzenetriol, 1,3,5-trihydroxybenzene, phloroglucin, phloroglucine, spasfon-lyoc, s-trihydroxybenzene, benzene-s-triol, 5-hydroxyresorcinol, benzene, trihydroxy
359
CHEBI:16204
benzene-1,3,5-triol
C1=C(C=C(C=C1O)O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 215.0°C to 218.0°C
Color White to Beige
Flash Point 100°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (HPLC)
Packaging Glass bottle
Linear Formula (OH)3C6H3
Quantity 500 g
Fieser 01,47; 06,147
Merck Index 15,7439
Solubility Information Solubility in water: 10g/L (20°C). Other solubilities: soluble in acetone ethanol, ether and pyridine
Formula Weight 126.1
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Phloroglucinol
Show More Show Less
compliance-icons
Product Identifier
  • Phloroglucinol, anhydrous
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'