missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphatase buffer 1 (5X), Thermo Scientific™
For cellular and molecular biology applications
37.00 € - 54.00 €
Specifications
Components | 100mM Tris (pH 7.5) and 100mM magnesium chloride |
---|---|
Content And Storage | Ambient temperatures |
Product Type | Phosphatase buffer 1 (5X) |
Form | Liquid |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15474669
|
Thermo Scientific Alfa Aesar
J62267.AK |
250 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15464669
|
Thermo Scientific Alfa Aesar
J62267.AE |
100 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Formulation: 100mM Tris (pH 7.5) and 100mM magnesium chloride
Specifications
100mM Tris (pH 7.5) and 100mM magnesium chloride | |
Ambient temperatures | |
Phosphatase buffer 1 (5X) | |
Liquid |
For Research Use Only