Learn More
PBS Tablets, Phosphate Buffered Saline, Fisher BioReagents™
Phosphate Buffered Saline Tablets, CAS Number-7647-14-5, 7447-40-7, 100 tablets, Clear, Gray, White, 5/8 in., 600 SEC max., CAUTION!
Brand: Fisher Bioreagents BP2944-100
137.80 EUR valid until 2024-03-30
Use promo code "21741" to get your promotional price.
Code : 32
Additional Details : CAS Number : 7447-40-7 Weight : 0.15000kg
Description
Phosphate Buffered Saline Tablets, CAS Number-7647-14-5, 7447-40-7, 100 tablets, Clear, Gray, White, 5/8 in., 600 SEC max., CAUTION!
- Save time and simplify your buffer preparation step by using Fisher BioReagents PBS tablets
- Eliminate the hassle of weighing and mixing individual components
- One tablet dissolved in 200mL water yields buffer containing 0.01M phosphate, 0.0027M KCl, and 0.137M NaCl, pH 7.4 at 25°C
- Bottle contains 100 tablets
- Intended for laboratory research use only
Specifications
White | |
100 tablets | |
7.2 to 7.6 |
Poly Bottle | |
Phosphate Buffered Saline | |
Solid |