missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-Bim (Thr56, Thr116) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA564655
Additional Details : Weight : 0.01000kg
Description
Phospho-Bim (Thr56, 116) Polyclonal Antibody detects endogenous levels of Bim only when phosphorylated at Thr56, 116.
Phospho-Bim (Thr56, Thr116) Polyclonal antibody specifically detects Phospho-Bim (Thr56, Thr116) in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
Phospho-Bim (Thr56, Thr116) | |
Polyclonal | |
Unconjugated | |
BCL2L11 | |
1500006F24Rik; BAM; Bcl2 interacting mediator of cell death; bcl-2 interacting mediator of cell death; bcl-2 interacting protein Bim; BCL2 like 11; Bcl-2 like 11; Bcl-2 related apoptotic gene product BimL; bcl2-interacting mediator of cell death; BCL2L11; Bcl2-L-11; BCL2-like 11 (apoptosis facilitator); bcl-2-like protein 11; bcl-2-like protein 11-like; bcl-2-related ovarian death agonist; bcl-2-related ovarian death protein; BIM; Bim EL; Bim-A; BIM-alpha6; BIM-beta6; BIM-beta7; BIMEL; BimEL protein; BimL; BOD; OTTHUMP00000197013; OTTHUMP00000204247; Unknown (protein for MGC:134444) | |
Rabbit | |
Affinity chromatography | |
RUO | |
10018, 12125, 64547 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O43521, O54918, O88498 | |
BCL2L11 | |
A synthesized peptide derived from human Bim (Phospho-Thr56/116) | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |