missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-TRPV1 (Ser502) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA564860
Additional Details : Weight : 0.01000kg
Description
Phospho-TRPV1 (Ser502) Polyclonal Antibody detects endogenous levels of TRPV1 only when phosphorylated at Ser502.
Phospho-TRPV1 (Ser502) Polyclonal antibody specifically detects Phospho-TRPV1 (Ser502) in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
Phospho-TRPV1 (Ser502) | |
Polyclonal | |
Unconjugated | |
TRPV1 | |
Capsaicin receptor; cation channel; DKFZp434K0220; non-selective cation channel; osm-9-like TRP channel 1; OTRPC1; si:dkey-9e15.2; transient receptor potential cation channel subfamily V member 1; transient receptor potential cation channel V1; transient receptor potential cation channel, subfamily V, member 1; transient receptor potential V1; transient receptor potential V1 channel; transient receptor potential vanilloid 1 supraoptic nucleus; transient receptor potential vanilloid 1a; transient receptor potential vanilloid 1b; TRPV1; TRPV1/2; TRPV1_SON; TRPV1alpha; TRPV1beta; Vanilloid receptor; vanilloid receptor 1; vanilloid receptor subtype 1; vanilloid receptor type 1 like protein 1; Vanilloid receptor type 1-like; vanilloid type 1 receptor; VR.5'sv; Vr1; VR-1; Vr1l; Vr1l1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
193034, 7442, 83810 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O35433, Q704Y3, Q8NER1 | |
TRPV1 | |
A synthesized peptide derived from human TRPV1 (Phospho-Ser502) | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |