missing translation for 'onlineSavingsMsg'
Learn More

Phosphotungstic Acid, 44-hydrate, ACROS Organics™

Brand:  Acros Organics 417931000

Code : 29

Additional Details : CAS Number : 12067-99-1 Weight : 0.15000kg Transport : UN number : 3260 Chem class : 8 Pack group : III


Product Code. 10005720

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 01 885 5854  for assistance.


Specifications

Specifications

Phosphotungstic acid, 44-hydrate
White to pale yellow
50ppm max.
Crystalline Powder
P2O5·24WO3·44H2O
300ppm max.
PTA
0.02% max.
50ppm max.
200ppm max.
6498.96
100g
12067-99-1
24WO3·44H2O
MFCD00149916
15, 7476
50ppm max.
30ppm max.
Glass bottle
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Phosphotungstic acid, 44-hydrate

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS