missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ Photometer Cell

Lovibond Photometer Cell, Height: 48mm, Includes: With Lid, Quantity: 5Pack

Brand:  Lovibond™ 197629

Additional Details : Weight : 0.09100kg

Product Code. 13576670

  • 39.00 € / Pack of 5
Estimated Shipment: 15-05-2024
to see stock.
Add to basket

Description

Description

Lovibond Photometer Cell, Height: 48mm, Includes: With Lid, Quantity: 5Pack

Specifications

Specifications

Photometer Cell
With Lid
5Pack
48 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers