missing translation for 'onlineSavingsMsg'
Learn More

Phthalic Anhydride, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog Number 15547804
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
85-44-9
C8H4O3
148.12
MFCD00005918
LGRFSURHDFAFJT-UHFFFAOYSA-N
phthalic anhydride, isobenzofuran-1,3-dione, 1,3-isobenzofurandione, 1,3-dioxophthalan, 1,3-phthalandione, phthalsaeureanhydrid, phthalic acid anhydride, o-phthalic acid anhydride, phthalandione, retarder esen
6811
CHEBI:36605
O=C1OC(=O)C2=CC=CC=C12
This item is not returnable. View return policy
85-44-9
C8H4O3
148.12
MFCD00005918
LGRFSURHDFAFJT-UHFFFAOYSA-N
phthalic anhydride, isobenzofuran-1,3-dione, 1,3-isobenzofurandione, 1,3-dioxophthalan, 1,3-phthalandione, phthalsaeureanhydrid, phthalic acid anhydride, o-phthalic acid anhydride, phthalandione, retarder esen
6811
CHEBI:36605
O=C1OC(=O)C2=CC=CC=C12

High quality chemicals for laboratory and specialized industrial use

Flash Point 152.00°C
Packaging Poly Bottle
Quantity 500 g
Specific Gravity 1L = 1.53kg
IUPAC Name 1,3-dihydro-2-benzofuran-1,3-dione
Formula Weight 148.12
Grade BAKER ANALYZED™ ACS Reagent
Chemical Name or Material Phthalic Anhydride
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'