Learn More
Thermo Scientific™ Pierce™ Top 12 Abundant Protein Depletion Spin Columns
Decrease the abundant albumin and antibody components of human plasma samples in preparation for mass spectrometry or 1D and 2D gel electrophoresis.
268.00 € - 898.00 €
Specifications
Description | Pierce Top 12 Abundant Protein Depletion Spin Columns |
---|---|
For Use With (Equipment) | Microcentrifuge, Mass Spectrometer |
Product Type | Spin Column |
Product Line | Pierce™ |
Content And Storage | Store at 4°C—do not freeze |
Product Code | Brand | Description | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Quantity | Price | Quantity & Availability | ||||
13434319
|
Thermo Scientific™
85165 |
Pierce Top 12 Abundant Protein Depletion Spin Columns | 24 Columns |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
13424319
|
Thermo Scientific™
85164 |
Pierce Top 12 Abundant Protein Depletion Spin Columns | 6 Columns |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Decrease the abundant albumin and antibody components of human plasma samples in preparation for mass spectrometry or 1D and 2D gel electrophoresis with Thermo Scientific™ Pierce Abundant Protein Depletion Spin Columns.
Additional Information and Online Ordering »
Highlights:
Optimized – resin in spin columns is scaled and optimized for treating 10μL (600μg) of human plasma samples for mass spectrometry and/or 1D and 2D electrophoresis
Efficient – kits remove >90% of IgG and >95% of albumin, plus other abundant proteins (Top 12)
Fast – process samples in 40 to 60 minutes (depending on resin)
Economical – cost effective spin columns are priced for single use
Reduce variability – One time use prevents abundant protein carryover and experimental variability
Two resin options – choose the system appropriate for your need: 2 or 12 protein depletion columns
Top 2 Spin Columns Remove:
- Albumin
- IgG
Top 12 Spin Columns Remove:
- Albumin
- IgG
- α1-Acid Glycoprotein
- α1-Antitrypsin
- α2-Macroglobulin
- Apolipoprotein A-I
- Apolipoprotein A-II
- Fibrinogen
- Haptoglobin
- IgA
- IgM
- Transferrin
Specifications
Pierce Top 12 Abundant Protein Depletion Spin Columns | |
Spin Column | |
Store at 4°C—do not freeze | |
Protein | |
Sample Enrichment, Sample Clean Up |
Microcentrifuge, Mass Spectrometer | |
Pierce™ | |
Plasma, Serum | |
Mass Spectrometry |