Learn More
Thermo Scientific™ CHCA MALDI Matrix, Single-Use
Highly purified, recrystallized CHCA, DHB and SA reagents in single-use tubes for mass spectrometry by matrix-assisted laser desorption ionization (MALDI).
Brand: Thermo Scientific™ 90031
246.69 EUR valid until 2024-03-29
Use promo code "21653" to get your promotional price.
Code : A0
Additional Details : Weight : 0.10000kg
Description
Thermo Scientific™ MALDI Matrices are highly purified, recrystallized reagents packaged in convenient single-use tubes sufficient for analyzing up to 96 samples by matrix assisted laser desorption ionization.
The MALDI method for MS peptide analysis is now easier to perform with single-use, highly pure MALDI matrices that are exceptionally convenient to use. Matrix assisted laser desorption ionization (MALDI) matrices produce the cleanest mass spectrometric (MS) spectra when they are recrystallized and prepared daily, which is inconvenient and typically results in significant waste. The packaging of purified, recrystallized alpha-cyano-4-hydroxy-cinnamic acid (CHCA), sinapinic acid (SA) and 2,5-dihydroxybenzoic acid (DHB) MALDI matrices makes preparation of high-quality MALDI reagents easy.Highlights:
- Highly purified, recrystallized reagent
- Packaged in convenient single-use tubes sufficient for analyzing up to 96 samples to save time and reduce waste
- Provides accurate quantity of matrix every time
Specifications
Pierce CHCA MALDI Matrix | |
MALDI Matrix | |
Small Molecules | |
Mass Spectrometry | |
Solid |
Mass Spectrometer | |
Upon receipt store product at 4°C. | |
Small Molecules | |
Calibration |
Safety and Handling
- CHCA MALDI Matrix, Single-Use
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : 2-cyano-3- (4-hydroxyphenyl) acrylic acid
For Research Use Only. Not for use in diagnostic procedures.