Learn More
Thermo Scientific™ Pierce™ Protease Inhibitor Tablets, EDTA-free
Thermo Scientific™ Pierce Protease Inhibitor Tablets are broad-spectrum protease inhibitors that are highly effective at preventing proteolytic degradation for cell lysis and protein extraction experiments
Brand: Thermo Scientific™ A32964
Additional Details : Weight : 0.03000kg
Alert:
For Research Use Only. Not for use in diagnostic procedures.
Description
Pierce Protease Inhibitor Tablets protect proteins from proteolytic degradation through inhibition of the serine-proteases, cysteine-proteases, aspartic acid-proteases and aminopeptidases that are typically present in cellular lysate samples. The tablets contain AEBSF, aprotinin, bestatin, E-64, leupeptin and pepstatin A. Stable for storage at 4°C for up to 12 months, each mini or large tablet tablet is sufficient for 10mL or 50mL of solution, respectively, and is suitable for reagent preparation prior to protein extraction from tissue and cultured cells.
Highlights:Specifications
1 vial of inhibitor tablets | |
protein purification, western blotting | |
2 Tablets |
2°C to 8°C | |
1 Tablet is Sufficient for 50 mL |
For Research Use Only. Not for use in diagnostic procedures.