missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Protein A/G Magnetic Beads

Product Code. 11814544 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
Unit Size:
1mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11814544 5 mL 5mL
11844554 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11814544 Supplier Thermo Scientific™ Supplier No. 88803
Save Up to 60% on Microbiology Essentials
Use promo code 28004
View all promotions
 Restricted Item
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Immunoprecipitate or affinity purify antibodies (IgG) using Protein A/G coated magnetic particles for manual or robotic magnetic separation.

Thermo Scientific Pierce Protein A/G Magnetic Beads are high-performance affinity particles for antibody purification and immunoprecipitation methods using manual or robotic magnetic separators.

Features of Protein A/G Magnetic Beads:

  • High capacity—nearly four times higher binding capacity than typical magnetic beads from other suppliers, allowing the use of smaller amounts per experiment
  • Low non-specific binding—stable, pre-blocked beads provide clean purification products (e.g., antigen eluted in IP with antibody is devoid of contaminating proteins from complex IP matrix)
  • Flexibility—convenience of IgG binding domains of both Protein A and Protein G on one bead
  • Compatibility—beads are compatible with manual and automated applications (e.g., Thermo Scientific KingFisher Instruments)
  • Assay consistency—magnetic beads eliminate resin loss and provide for more efficient separation of solutions than traditional IP methods that use only microcentrifuge tubes

These magnetic beads are coated with genetically engineered Pierce Protein A/G, a recombinant fusion protein which combines the IgG binding domains of both Protein A and Protein G. This enables capture of antibodies from a wider range of species and isotypes than either protein alone. Using our crosslinker chemistry, you can immobilize an antibody onto the magnetic particle and prevent IgG contamination in your immunoprecipitated sample. These beads can be used both manually with a magnetic stand as well as with automated platforms such as the Thermo Scientific KingFisher Instruments.

Applications:

  • IP and Co-IP experiments (see complete kit)
  • Immunoprecipitation for analysis in non-reducing conditions
  • Antibody purification

The recombinant Protein A/G that is immobilized onto the Pierce Magnetic Beads is a fusion of the IgG binding domains of both Protein A and Protein G. Protein A/G contains four Fc-binding domains from Protein A and two from Protein G, making it a convenient tool for investigating and purifying immunoglobulins. Thus, Pierce Magnetic Particles are not simply a mixed immobilization of separate Protein A and Protein G polypeptides, nor are they a mixture of Protein A magnetic beads and Protein G magnetic beads.

TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 5 mL
Concentration Slurry: 10 mg/mL, 1% solids
Content And Storage Upon receipt store at 4°C. Product shipped with an ice pack.
Particle Size 1 μm
Form Liquid
Product Line Pierce
Quantity 5 mL
Protein Form Recombinant
Target Antibody
Type Magnetic Affinity Bead
Ligand Type Protein A/G
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.