missing translation for 'onlineSavingsMsg'
Learn More

ROBU VitraPOR™ Pipeline Filter

Pipeline Filter with tube adapters for connection.

Brand:  ROBU VitraPOR™ 23601

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10683752

  • 94.20 € / Each
Estimated Shipment: 24-06-2024
to see stock.
Add to basket

Description

Description

  • Highly resistant to most chemicals and temperatures up to 500°C
Specifications

Specifications

Borosilicate Glass (3.3)
Non-sterile
1 Ea.
140 mm
1 μm
Tube Adapters
1
60 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers