missing translation for 'onlineSavingsMsg'
Learn More

ROBU VitraPOR™ Pipeline Filter

Pipeline Filter with tube adapters for connection.

Brand:  ROBU VitraPOR™ 23300

Additional Details : Weight : 0.09120kg

 View more versions of this product

Product Code. 16863046

  • 83.15 € / Pack of 2
Estimated Shipment: 30-05-2024
to see stock.
Add to basket

Description

Description

  • Highly resistant to most chemicals and temperatures up to 500°C
Specifications

Specifications

Borosilicate Glass (3.3)
Tube Adapters
0
30 mm
Non-sterile
2/Pk.
140 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers