missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manutan Platform trolley, steel, foldable, 150kg capacity
Brand: Manutan A340336
Code : Z3
Additional Details : Weight : 1.00000kg
Description
Highly durable welded steel trolleys with a foldable handle. Non-slip PVC platform surface, ensures maximum grip of load when in transit. All round buffer strip protects walls and door frames when the truck is in use. Welded steel construction.- Dimensions (W x L): 480mm x 740mm
- Handle height: 920mm
- Load capacity: 150kg
- Weight: 12.5kg
- Foot operated handle release bar
- Two fixed and two rear swivel non-marking tyres.