missing translation for 'onlineSavingsMsg'
Learn More

Platinum Cobalt Color Standard Solution (APHA), Fisher Chemical™

Brand:  Fisher Bioreagents USSP120500

Code : NEW

Additional Details : CAS Number : 7647-01-0 Weight : 0.50000kg Transport : UN number : 1789 Chem class : 8 Pack group : III

 View more versions of this product

Product Code. 11987175

  • 87.10 € / 500mL
Estimated Shipment: 07-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Yellow
Liquid
500 mL
1.1
Glass Bottle
Platinum Cobalt Color Standard Solution
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Special Offers

Special Offers