Learn More
Thermo Scientific™ Plunger-in-Needle Autosampler Syringes for Thermo Scientific™ Instruments
Get accurate and reproducible results over a range of volumes with Thermo Scientific™ Plunger-in-Needle Autosampler Syringes for Thermo Scientific Instruments.
133.62 € - 159.10 €
Specifications
Disposable No
Volume (Metric) 0.5 μL
Needle Point Style Cone
View More Specs
Products 3
Product Code Brand Needle Gauge Price Quantity & Availability  
Product Code Brand Needle Gauge Price Quantity & Availability  
12886036
View Documents Promotion Details
Thermo Scientific™
365A0241
23 G
159.10 €
Each
View alternative products
Save Up to 
»
Estimated Shipment: 21-05-2024
to see stock.
10585245
View Documents Promotion Details
Thermo Scientific™
36504046
26 G
142.30 €
Each
View alternative products
Save Up to 
»
Estimated Shipment: 01-05-2024
to see stock.
10310975
View Documents Promotion Details
Thermo Scientific™
36500500
31 G This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description
Description
Get accurate and reproducible results over a range of volumes with Thermo Scientific™ Plunger-in-Needle Autosampler Syringes for Thermo Scientific Instruments.
  • Manual syringes in industry-standard size
  • White scale backing for easy reading
  • Solvent-resistant markings
Specifications
Specifications
No
0.5 μL
Cone