missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ PolarSafe™ Cryogenic Storage and Transport Dewars

Cole-Parmer™ PolarSafe™ Cryogenic Storage and Transport Dewars are robust cryopreservation solutions for long-term storage and transport of biological samples

Brand:  Cole-Parmer™ 4426065

Additional Details : Weight : 12.24700kg

 View more versions of this product

Product Code. 16609561

  • 835.00 € / Each
Estimated Shipment: 20-12-2024
to see stock.
Add to basket

Description

Description

  • Strong yet lightweight aluminum dewars hold liquid nitrogen
  • Narrow neck design provides high thermal efficiency and low evaporation loss
  • Heavy duty lockable enclosure for enhanced security
  • Long term storage up to 200 days (model dependent)
  • Seven sizes available from 2 L to 35 L
  • 5 year vacuum warranty
Specifications

Specifications

5 cm
2 in.
Static Holding Time: 126 Days
Aluminum
43.18 cm
43.18 cm
17 in.
17 in.
20 L
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers