Learn More
Cole-Parmer™ PolarSafe™ Stainless Steel Liquid Nitrogen Refill Tank
Cole-Parmer™ PolarSafe™ Stainless Steel Liquid Nitrogen Refill Tank, with self-pressurized liquid nitrogen, makes LN2 discharge safe and simple
Brand: Cole-Parmer™ 4426069
Additional Details : Weight : 59.87420kg
Description
- Durable welded stainless steel construction ensures long-lasting performance and reduced evaporation loss
- Unique design utilizes pressure from LN2 to discharge LN2 to other containers
- Use as a self-pressurized LN2 backup system for cryogenic freezers
- 5 year vacuum warranty
Specifications
50.546 cm | |
19.9 in. | |
25 in. | |
50 L |
63.5 cm | |
25 in. | |
Stainless Steel | |
63.5 cm |