Learn More
Abnova™ POLR3F 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Brand:  Abnova™ H00010621-T01.100uL
Additional Details : Weight : 0.00010kg
Product Code. 16162916
  • 209.00 € / 100µL
Estimated Shipment: 04-06-2024
to see stock.
Specifications
Western Blot
POLR3F 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 34.87kDa
Transient overexpression cell lysate was tested with anti-POLR3F antibody (Mfr. No. H00010621-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
293T
NM_006466.2
pCMV-POLR3F full-length