Learn More
Invitrogen™ Poly(A) Tail-Length Assay Kit
Newly reconfigured kit for increased efficiency.
Brand: Invitrogen™ 764551KT
Additional Details : Weight : 0.40000kg
Description
- Accurate measurement of poly(A) tail length compared to other assays (e.g. northern blotting)\
- One kit includes optimized reagents for 5 G/I Tailing, 20 RT and 80 PCR reactions
- Poly adenylation of eukaryotic mRNAs plays a critical role in RNA metabolism including stability, enhanced translation, nuclear export and miRNA mediated gene regulation.
- Allows you to quickly measure the poly(A) tails of multiple mRNAs in only three hours.
- This kit utilizes a novel G/I extension technique which preserves the 3' end of RNAs and ensures accurate measurement of poly(A) tail-length. And, all reaction components have been optimized so you get quality data quickly.
- Kit contains all necessary reagents for 5 G/I Tailing, 20 reverse transcription, and 80 PCR reactions
Specifications
Measurement of poly(A) tail-lengths | |
Poly(A) RNA Polymerase | |
Poly(A) tail-length detection | |
100 ng to 2 μg of total eukaryotic RNA |
For Research Use Only. Not for use in diagnostic procedures.