missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manutan Polycarbonate blue door cabinet with three shelves,1830mm x 915mm x 457mm
Brand: Manutan A343108
Code : Z3
Additional Details : Weight : 1.00000kg
Description
Strong welded construction. Floor Standing Cabinet- Blue door
- Three shelves
- Dimensions (H x W x D): 1830mm x 915mm x 457mm