Learn More
Masterflex™ Polycarbonate Luer Fittings
Designed to work with microtubing and luer-type syringes. Standard-sized connecting ends make these luer fittings completely interchangeable for surefire installations.
Brand: Masterflex™ 45504-04
Code : A0
Additional Details : Weight : 0.01620kg
Description
Designed to work with microtubing and luer-type syringes. Standard-sized connecting ends make these luer fittings completely interchangeable for surefire installations.
- Manufactured from 100% virgin raw materials
- Free of contaminants and ideal for syringes or small tubing
- Choose from a range of sizes with a variety of connection options
- Easily connect fluid paths without the use of tools
- Male and female luers available
Specifications
Luer Fitting | |
Polycarbonate | |
For flexible tubing, reduce flow turbulence, semi-rigid tubing |
ISO 9001 | |
Male luer with lock ring x 1/8 in. hose barb | |
25 Pack |