Learn More
Masterflex™ Polycarbonate Luer Fittings
Designed to work with microtubing and luer-type syringes. Standard-sized connecting ends make these luer fittings completely interchangeable for surefire installations.
Brand:  Masterflex™ 45504-00
Code : A0
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 12197750
  • 24.30 € / Pack of 25
Estimated Shipment: 23-07-2024
to see stock.
Add to basket
Description
Description
Designed to work with microtubing and luer-type syringes. Standard-sized connecting ends make these luer fittings completely interchangeable for surefire installations.
  • Manufactured from 100% virgin raw materials
  • Free of contaminants and ideal for syringes or small tubing
  • Choose from a range of sizes with a variety of connection options
  • Easily connect fluid paths without the use of tools
  • Male and female luers available
Specifications
Specifications
Luer Fitting
Polycarbonate
For flexible tubing, reduce flow turbulence, semi-rigid tubing
ISO 9001
Male luer with lock ring x 1/16 in. hose barb
25 Pack