Learn More
Macherey-Nagel™ Polyester Sheets Polygram SIL G, Alox N, CEL 300/UV254 TLC Micro-Set M
Brand:  Macherey-Nagel™ 814028
Code : NEW
Additional Details : Weight : 0.01000kg
Product Code. 10125141
  • 149.00 € / Pack of 200
Estimated Shipment: 23-05-2024
to see stock.
Add to basket
Specifications
Polyester Sheets Polygram SIL G, Alox N, CEL 300
TLC Micro-Set M
2 developing chambers; 2 x 50 glass capillaries 1 μL; 1 rubber cap for glass capillaries; 2 spotting guides; 1 measuring cylinder 10 mL; 1 beaker 25 mL; 1 glass laboratory sprayer with rubber bulb; 1 plastic syringe 1 mL; 20 Sheets filter paper MN 713, 15 x 21 cm; 50 polyester sheets Polygram SIL G/UV254, Alox N/UV254, CEL 300
200 Pk.
UV254
4 x 8 cm