Learn More
Macherey-Nagel™ Polygram CEL 300 Polyester Sheets
TLC polyester sheets with native fibrous cellulose. Fiber length (95 %) 2–20 μm, average degree of polymerization 400–500, specific surface acc. to Blaine 15000 cm2/g; ≤ 20 ppm Fe, 6 ppm Cu, 7 ppm P; dichloromethane extract ≤ 0.25 %; residue on ignition at 850 °C ≤ 1500 ppm.
Brand: Macherey-Nagel™ 801011
Code : 50
Additional Details : Weight : 0.01000kg
Description
TLC Polyester sheets, cellulose layer, POLYGRAM CEL 300
- Base material: Polyester sheets coated in cellulose
- Mode: Normal Phase
- Layer Hardness: Standard, Soft
Adsorbent:
- Polyester sheets coated in cellulose
- Phase CEL 300
- UV Indicator: No
- Particle Type: Fully porous particles (FPP)
- Particle Shape: Irregular
- Particle size 2-20 μm
- Binder: Yes
Indicators:
This plate can be used different UV indicators - manganese activated zinc silicate with green fluorescence for short-wave UV (254 nm); special inorganic fluorescent pigment with blue fluorescence for long-wave UV (366 nm).
Binder:
Highly polymeric products, which are stable in almost all organic solvents and resistant towards aggressive visualisation reagents
Recommended Applications:
Amino acids, Carbohydrates, Carboxylic acids, Polar substances
General TLC Glass Plate Features from Macherey-Nagel:
- Pre-coated ready to use plates
- Broad range of plate sizes, surface coatings and backings
- The plate can be easily labelled for clear identification
- Glass plates are robust, heat proof and chemically resistant to all common mobile phases and visualization reagents.
- Increased separation efficiency due to an optimized particle size distribution
- A dust free plate allows convenient handling and avoids silica contamination of the TLC chamber
Specifications
50 Pk. | |
Polyester Sheet | |
0.1 mm | |
4 x 8 cm |