missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Polygram POLYAMIDE-6 Polyester Sheets
134.00 € - 221.00 €
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
10496661
|
Macherey-Nagel™
803013 |
20 x 20 cm |
221.00 €
Pack of 25 |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||||
10699961
|
Macherey-Nagel™
803012 |
5 x 20 cm |
134.00 €
Pack of 50 |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||||
Specifications
Polyester Sheet | |
0.1 mm |