Learn More
Kartell™ Polyoxymethylene Clamps
Manufactured from polyoxymethylene for low friction and excellent dimensional stability. Kartell™ Polyoxymethylene Clamps are color coded according to joint size and provide a safe connection for glass conical joints.
Brand:  Kartell™ 3293200
Code : A0
Additional Details : Weight : 0.00940kg
 View more versions of this product
Product Code. 10428072
  • 3.30 € / Each
UK Stock 1
Add to basket
You may only order up to the available stock. For more details call customer services on 01 885 5854 .
Description
Description
  • Plastic construction generates a corrosion proof device that does not scratch glass
  • Available in various colors that correspond with the joint size of the connection
  • Autoclavable
  • Safe connecction for standard-finish glass conical joints
Specifications
Specifications
Autoclavable
Red
Clamp
Polyoxymethylene
Standard-finish glass conical joint size 29/32
1Pack
Videos