missing translation for 'onlineSavingsMsg'
Learn More

Polyphosphoric Acid, Reagent grade, 115% H3PO4 basis, Honeywell™

Catalog Number p-7120675
Click to view available options
Quantity:
25 g
1 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
8017-16-1
(HO3P)n
MFCD00084480
NBIIXXVUZAFLBC-UHFFFAOYSA-N
Polyphosphoric acid
[H]OP(O)(O)=O
This item is not returnable. View return policy
8017-16-1
(HO3P)n
MFCD00084480
NBIIXXVUZAFLBC-UHFFFAOYSA-N
Polyphosphoric acid
[H]OP(O)(O)=O

Reagent Grade, 115% H3PO4 basis

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY
Color Colorless
Boiling Point 300°C
Density 2.06g/mL at 25°C
Packaging Poly Bottle
Quantity 25 g
Linear Formula Hn+2PnO3n+1
Physical Form Liquid
UN Number UN3264
Percent Purity 115% (H3PO4 basis)
Grade Reagent
Chemical Name or Material Polyphosphoric Acid
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'