Learn More
BRAND™ Polypropylene Cap Inserts
Providing a secure fit for all sizes.
Brand: BRAND™ 114852
20.47 EUR valid until 2024-12-31
Use promo code "21935" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.07000kg
Description
- Available in different colors
- Provided in packs of 500
- High temperature and chemical resistance
Specifications
Red | |
Polypropylene | |
500/Pk. |