missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Chromabond™ XTR Adsorbent
Use Chromabond™ XTR with Kieselguhr phase for liquid-liquid extraction. Chromabond™ XTR Adsorbent provides options for Chromabond™ XTR columns.
Brand: Macherey-Nagel™ 730586
Code : 50
Additional Details : Weight : 0.01000kg
Description
Features- Base material coarse-grained kieselguhr (also known as diatomaceous earth, hydromatrix, celite)
- large pore size, high pore volume, constantly high batch-to-batch quality
- pH working range 1–13
- Liquid-liquid extraction of highly viscous aqueous solutions such as physiological fluids (blood, plasma, and serum) in clinical chemistry, dyes in textiles, environmental and food analysis without use of a separation funnel
- High water loadability without breakthrough of water during elution with organic solvents
- Also suited for removing small amounts of water from solvents which are not miscible with water
Advantages
- Fast, reproducible and economical
- Simultaneous preparation of several samples
- No problems with phase separation ∙ no formation of emulsions
- High recovery rates
- Saving of time and solvents
- Organic solutions need not to be dried after separation
Solvents applicable for elution
- diethyl ether
- tert-butyl methyl ether
- ethyl acetate
- n-hexane
- cyclohexane
- toluene
- dichloromethane (methylene chloride)
- trichloromethane (chloroform)
- solvent mixtures:
- trichloromethane – methanol (90:10, v/v)
- trichloromethane – methanol (85:15, v/v)
- diethyl ether – ethanol (90:10, v/v)
- diethyl ether – ethanol (80:20, v/v)
- dichloromethane – 2-propanol (90:10, v/v)
- dichloromethane – 2-propanol (85:15, v/v)
Specifications
50 Pack | |
NT20 | |
XTR Adsorbent | |
PE filter elements |