missing translation for 'onlineSavingsMsg'
Learn More

Ratiolab™ Polypropylene Grid Inserts

Brand:  Ratiolab™ 5220101

Additional Details : Weight : 100.00000kg

 View more versions of this product

Product Code. 15951651

  • 15.35 € / Pack of 5
Estimated Shipment: 13-08-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

Polypropylene
Insert Grid
5 Pack
133 mm
Square
17.3 mm Diameter Tubes
133 mm
30 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers