missing translation for 'onlineSavingsMsg'
Learn More

Merck Millipore Polypropylene Microfil Funnel & Filter

Convenient, easy to use Microfil™ membrane funnel and filteration system

Brand:  Merck Millipore MIGSWG250

Code : NEW

Additional Details : Weight : 0.03000kg Transport : UN number : 3270 Chem class : 4.1 Pack group : II


 View more versions of this product

Product Code. 11953866

Quantity Price
1 367.00 € / Pack of 150
6 320.50 € / Pack of 150
Estimated Shipment
19-07-2021
Add to basket


Description

Description

  • Convenient, easy to use system for well accepted membrane filtration method conforms to international standards: CEN, Standard Methods, WHO
  • Disposable, stackable, light, recyclable plastic Microfil™ funnels eliminate autoclaving and minimise waste
  • 'Push fit' Microfil™ funnels tightly seal to manifold supports without clamps or O-rings assuring leak-free operation and uniform micro-organism recoveries
  • Novel device lifts membrane after filtration, for easy removal with forceps
  • Unique Microfil™ support filters faster than all-glass filter apparatus
  • 100mL or 250mL funnels available with convenient accessory dispensers
  • Choose conventional sterile membranes or novel EZ-Pak™ membrane cartridges. Select manifold-mounted filtration supports for laboratory use or stand-alone on-site filtration support.
Specifications

Specifications

White
Specific test regulations, Clinical Lab, Food and Beverage, Industrial QC , environmental Analysis, Laboratory Filtration
Microfil Funnel and Filter
0.22 μm
47 mm
150/Pk.
Polypropylene
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Microfil Funnel and Filter, 250 ml, 0,22

Signal Word
  • Danger

Hazard Category
  • Flammable solid Category 1

Hazard Statement
  • H228-Flammable solid.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P370+P378-In case of fire: Use to extinguish.

Supplemental information
  • MIXTURE LIST-Contains : Octylphenol polyethoxyethanol

SDS
Documents