missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VITLAB™ Polypropylene Standard Joint Funnels
Fill multi-neck reaction flasks with standard neck joints. VITLAB™ Polypropylene Standard Joint Funnels are transparent, laterally flattened funnels that fit snugly into standard neck joints for fast, easy filling. Available for a variety of standard neck sizes.
POA
Specifications
Product Type | Standard Joint Funnel |
---|---|
Material | Polypropylene |
Color | Transparent |
For Use With (Application) | Suitable for the Filling of Liquid or Powdered Reagents Into a Reaction Flask During a Reaction |
For Use With (Equipment) | Multi-Neck Flasks and Single-Neck Flasks with Standard Joint Necks |
Description
Suitable for the filling of liquid or powdered reagents into a reaction flask during a reaction- Laterally flattened for use with multi-neck flasks
- Suitable for standard joint necks of various sizes
- Transparent material for easy monitoring of filling progress
- Constructed of durable polypropylene
- Chemically resistant for use with liquid or powdered reagents
- Temperature resistant and autoclavable
- FDA-grade quality
Specifications
Standard Joint Funnel | |
Transparent | |
Multi-Neck Flasks and Single-Neck Flasks with Standard Joint Necks |
Polypropylene | |
Suitable for the Filling of Liquid or Powdered Reagents Into a Reaction Flask During a Reaction |