Learn More
Cole-Parmer™ Polypropylene Storage Container Lids
Use these lids for sample containers 06100-12 and 06100-14. Cole-Parmer™ Polypropylene Storage Container Lids and containers are chemical resistant to acids bases and most organic solvents.
Brand: Cole-Parmer™ 06100-16
Code : A0
Additional Details : Weight : 2.78100kg
Description
- 500 lids per case
- Made of polypropylene (PP)
- Resist most chemicals
- Containers sold separately
Specifications
500/CS | |
Storage Container Lid | |
Polypropylene | |
Sample Containers 06100-12 and 06100-14 |