missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kartell™ Plastilab™ Polypropylene Y-Shaped Untapered Connectors
For use with any hose system and vacuum lines.
Brand: Kartell™ 7052500
Code : A0
Additional Details : Weight : 0.05400kg
Description
- For use with hose systems and vacuum lines
Specifications
CE IVD | |
14 mm | |
Polypropylene | |
For any hoses system and vacuum line | |
9.7mm Bore |
Natural | |
Autoclavable | |
Untapered Y Tubing Connector | |
10/Pk. | |
Y |