missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brand™ Polystyrene Closure Tube
Brand: Brand™ 114750
Code : NEW
Additional Details : Weight : 4.50000kg
Description
- Sample tube (coagulometer)
- 11 x 55 mm
Specifications
Sample Tube | |
55 mm | |
5000 Pack |
11 mm | |
Polystyrene |