missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LABPLAS Polyurethane Sani-Stick with Letheen Broth
Description
- Polyurethane Sani-Stick
- Saturated with Letheen broth
- Dimensions: 160 x 229 mm
- Holds: 600 mL
- Option to include gloves with kit
Specifications
Specifications
| Product Type | Hygiene Sponge Sampling Kit |
| Media Type | Neutralizers |
| Sterility | Sterile |
| Includes | None |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?