missing translation for 'onlineSavingsMsg'
Learn More

POPSO, 0.2M buffer soln., pH 8.0, Thermo Scientific™

Catalog Number p-7041742
Click to view available options
Quantity:
100 mL
250 mL
This item is not returnable. View return policy
108321-07-9
C10H20N2Na2O8S2
406.376
MFCD00070005
XMRCLLXGMGEVHS-UHFFFAOYSA-L
53393457
disodium;2-hydroxy-3-[4-(2-hydroxy-3-sulfonatopropyl)piperazin-1-yl]propane-1-sulfonate
C1CN(CCN1CC(CS(=O)(=O)[O-])O)CC(CS(=O)(=O)[O-])O.[Na+].[Na+]
This item is not returnable. View return policy
108321-07-9
C10H20N2Na2O8S2
406.376
MFCD00070005
XMRCLLXGMGEVHS-UHFFFAOYSA-L
53393457
disodium;2-hydroxy-3-[4-(2-hydroxy-3-sulfonatopropyl)piperazin-1-yl]propane-1-sulfonate
C1CN(CCN1CC(CS(=O)(=O)[O-])O)CC(CS(=O)(=O)[O-])O.[Na+].[Na+]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Concentration 0.2 M
pH 8.0
Physical Form Liquid
Quantity 100 mL
Chemical Name or Material POPSO buffer soln.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'