Learn More
Fisherbrand™ Porcelain Buchner Funnels
Excellent for separating liquids from solids in the laboratory. Fisherbrand™ Porcelain Buchner Funnels provide a quick and easy pressure assisted filtration method.
POA
Description
- White porcelain
- Available for a range of volumes and filter diameters
- Applications include collecting recrystalized compounds
- Excellent thermal and chemical resistance
Specifications
Buchner Funnel | |
Porcelain | |
1Each |