missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Haldenwanger™ Porcelain Medium Form Crucibles
Use these dishes for a variety of applications. Haldenwanger™ Porcelain Medium Form Crucibles feature a smooth internal finish, constructed from optimized laboratory porcelain.
15.30 € - 161.12 €
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
12316507
|
Haldenwanger™
79MF/2 |
150 mL |
161.12 €
Pack of 10 |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
12336497
|
Haldenwanger™
L 79MF/8 |
10 mL |
65.50 €
Pack of 10 |
In Stock
Log in to see stock. |
|||||
12386497
|
Haldenwanger™
79MF/5 |
60 mL |
92.64 €
Pack of 10 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
|||||
10539411
|
Haldenwanger™
L 79MF/2 |
150 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Available in a variety of sizes
- Sizes up to 175mL glazed except rim
- 1,000mL and larger are glazed except outside base
- Complies with DIN VDE 0335, type C 100
Specifications
Porcelain | |
Crucible |