missing translation for 'onlineSavingsMsg'
Learn More

Potassium (2-nitrophenyl)trifluoroborate, 97%, Thermo Scientific™

Catalog Number p-4380104
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
850623-64-2
C6H4BF3KNO2
229.01
DZHYOFKWMHZKBK-UHFFFAOYSA-N
potassium 2-nitrophenyl trifluoroborate, potassium trifluoro 2-nitrophenyl borate, potassium trifluoro 2-nitrophenyl boranuide, potassium trifluoro-2-nitrophenyl boranuide, potassium 2-nitrophenyltrifluoroborate, amtb936, potassium ion trifluoro 2-nitrophenyl boranuide, potassium tris fluoranyl-2-nitrophenyl boranuide
2782854
potassium;trifluoro-(2-nitrophenyl)boranuide
[B-](C1=CC=CC=C1[N+](=O)[O-])(F)(F)F.[K+]
This item is not returnable. View return policy
850623-64-2
C6H4BF3KNO2
229.01
DZHYOFKWMHZKBK-UHFFFAOYSA-N
potassium 2-nitrophenyl trifluoroborate, potassium trifluoro 2-nitrophenyl borate, potassium trifluoro 2-nitrophenyl boranuide, potassium trifluoro-2-nitrophenyl boranuide, potassium 2-nitrophenyltrifluoroborate, amtb936, potassium ion trifluoro 2-nitrophenyl boranuide, potassium tris fluoranyl-2-nitrophenyl boranuide
2782854
potassium;trifluoro-(2-nitrophenyl)boranuide
[B-](C1=CC=CC=C1[N+](=O)[O-])(F)(F)F.[K+]
Melting Point 226°C to 228°C
Assay Percent Range 97%
Packaging Glass Bottle
Quantity 1 g
Formula Weight 229.01
Percent Purity 97%
Chemical Name or Material Potassium (2-nitrophenyl)trifluoroborate
compliance-icons
Product Identifier
  • Potassium (2-nitrophenyl)trifluoroborate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'