missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium 4-chlorophenyltrifluoroborate, 96%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Potassium 4-chlorophenyltrifluoroborate is involved in Suzuki Cross-Coupling reaction as potent boronic acid surrogates.
Solubility
Slightly soluble in water.
Notes
Incompatible with oxidizing agents and acids. Store at room temperature.
Potassium 4-chlorophenyltrifluoroborate is involved in Suzuki Cross-Coupling reaction as potent boronic acid surrogates.
Solubility
Slightly soluble in water.
Notes
Incompatible with oxidizing agents and acids. Store at room temperature.
Specifications
Melting Point | 293°C to 295°C |
Solubility Information | Slightly soluble in water. |
Formula Weight | 218.45 |
Percent Purity | 96% |
Quantity | 1 g |
Chemical Name or Material | Potassium 4-chlorophenyltrifluoroborate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'