missing translation for 'onlineSavingsMsg'
Learn More

Potassium bromide, FT-IR Grade, ≥99% trace metals basis, Honeywell Fluka™

Catalog Number 15632160
Click to view available options
Quantity:
25 g
100 g
7758-02-3
BrK
119.00
MFCD00011358
IOLCXVTUBQKXJR-UHFFFAOYSA-M
potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
253877
CHEBI:32030
[K+].[Br-]
7758-02-3
BrK
119.00
MFCD00011358
IOLCXVTUBQKXJR-UHFFFAOYSA-M
potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
253877
CHEBI:32030
[K+].[Br-]
Quantity 100 g
Linear Formula KBr
UN Number NONH for all modes of transport
IUPAC Name potassium;bromide
Formula Weight 119g/mol
Percent Purity ≥99% (trace metals basis)
Packaging Poly bottle
Vapor Pressure 1mmHg (795°C), 0.01mmHg (20°C)
Chemical Name or Material Potassium Bromide
Product Identifier
  • Potassium bromide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'