missing translation for 'onlineSavingsMsg'
Learn More

Potassium carbonate, Puratronic™, 99.997% (metals basis), Thermo Scientific Chemicals

Catalog Number 15475965
Click to view available options
Quantity:
5 g
25 g
100 g
584-08-7
CK2O3
138.21
MFCD00011382
BWHMMNNQKKPAPP-UHFFFAOYSA-L
potassium carbonate, dipotassium carbonate, potash, potassium carbonate, anhydrous, carbonic acid, dipotassium salt, carbonate of potash, pearl ash, salt of tartar, kaliumcarbonat, potassiumcarbonate
11430
[K+].[K+].[O-]C([O-])=O
584-08-7
CK2O3
138.21
MFCD00011382
BWHMMNNQKKPAPP-UHFFFAOYSA-L
potassium carbonate, dipotassium carbonate, potash, potassium carbonate, anhydrous, carbonic acid, dipotassium salt, carbonate of potash, pearl ash, salt of tartar, kaliumcarbonat, potassiumcarbonate
11430
[K+].[K+].[O-]C([O-])=O

Potassium carbonate has a wide range of industrial and research applications. The major use of potassium carbonate is the manufacture of chemicals, fertilizers, soap and glass. It is used as a drying agent for solvents especially if there are traces of acids present, and as a scavenger of acids produced in organic synthesis. It is used as a buffering agent during the production of wine. It is also used as a fire suppressant in extinguishers. In organic synthesis, it is an extensively used reagent for the alkylation and arylation reactions on carbon, sulfur, nitrogen, and oxygen centres. It is also used in aldol condensation, Michael reaction, acylation, Wittig reaction, Knoevenagel reaction, elimination reaction, epoxide opening, cyclization, metal-catalyzed cross coupling reaction, hydration, and a wide array of reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 891°C
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.99% (metals basis)
Beilstein 4267587
Merck Index 14,7619
Solubility Information Freely soluble in water. Insoluble in acetone,and alcohol.
IUPAC Name dipotassium carbonate
Formula Weight 138.21
Percent Purity 99.997%
Grade Puratronic™
Sensitivity Hygroscopic
Density 2.29
Assay (metals basis)
Chemical Name or Material Potassium carbonate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'