missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Carbonate, Anhydrous, Granular, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Wide Mouth Amber Glass Bottle
Specifications
Physical Form | Granules |
Quantity | 500 g |
IUPAC Name | dipotassium carbonate |
Formula Weight | 138.21 |
Grade | BAKER ANALYZED™ ACS Reagent |
Packaging | Wide Mouth Amber Glass Bottle |
Specific Gravity | 1L = 2.29kg |
Chemical Name or Material | Potassium Carbonate, Anhydrous |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'