missing translation for 'onlineSavingsMsg'
Learn More

Potassium chloride, 0.00068M, Conductivity Standard, Thermo Scientific Chemicals

Catalog Number p-4782069
Click to view available options
Quantity:
500 mL
1 L
4 L
This item is not returnable. View return policy
7447-40-7, 7732-18-5
ClK
74.55
MFCD00011360
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
4873
CHEBI:32588
[Cl-].[K+]
This item is not returnable. View return policy
7447-40-7, 7732-18-5
ClK
74.55
MFCD00011360
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
4873
CHEBI:32588
[Cl-].[K+]

Potassium chloride 0.00068M solution is used as conductive standard and as electrolyte in analytical chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 500 mL
Color Colorless
Assay Percent Range 0.00068M
IUPAC Name potassium chloride
Formula Weight 74.55
Concentration or Composition (by Analyte or Components) Potassium chloride: 0.005%; Water: 99.995%
Odor Odorless
Vapor Pressure 17mm Hg at 20°C
Solubility Miscible with water
Physical Form Liquid
Chemical Name or Material Potassium chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'