Learn More
Potassium Chromate Indicator 5% Solution, Reagecon
High-quality indicator solution produced from the highest quality raw materials and manufactured and tested under rigorous conditions.
Brand: Reagecon PCS5
Code : NEW
Additional Details : CAS Number : 7789-00-6 Weight : 0.54400kg Transport : UN number : 3082 Chem class : 9 Pack group : III
Description
- Titrant, Standard Titrant, Standard Solution, Standardized Solution, Reference Standard, Reference Material
- For manual titrations; indicator titrant or analyte change color and this color change are by far the most important method of end point detection.
- This high-quality indicator solution facilitates end point detection, in a clear and unambiguous manner and is an imperative for accurate manual titrimetry.
Specifications
Accuracy ± 0.1% | |
HDPE Bottle GL45 | |
Titration & Analyte Color Change |
0.05 | |
500 mL | |
Liquid |
Safety and Handling
- potassium chromate solution 5%
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 2
- Germ cell mutagenicity Category 1B
- Carcinogenicity Category 1B
- Skin sensitiser Category 1
Hazard Statement
- H340-May cause genetic defects.
- H350i-May cause cancer by inhalation.
- H411-Toxic to aquatic life with long lasting effects.
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : potassium chromate